missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Urabrelimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Urabrelimab |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282848
|
Invitrogen™
MA559231 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284763
|
Invitrogen™
MA559232 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Urabrelimab, also known as SRF231, is an investigational human monoclonal antibody targeting the CD47 protein, a cell surface antigen involved in immune evasion by cancer cells. The CD47 gene encodes a transmembrane glycoprotein broadly expressed on the surface of various cell types. Functionally, CD47 serves as a 'marker of self' by binding to the Signal-regulatory protein alpha (SIRP alpha) receptor on myeloid cells, thereby preventing macrophage-mediated phagocytosis. This interaction is crucial in maintaining homeostasis by inhibiting the immune system from attacking healthy cells. However, CD47 is significantly overexpressed on the surface of many tumor cells, providing a mechanism for these cells to evade the immune system. As such, CD47 is a prominent target in cancer immunotherapy aimed at blocking this 'don't eat me' signal to enhance the phagocytosis of cancer cells and improve patient prognosis.Specifications
| Urabrelimab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG4SP | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title