Learn More
Invitrogen™ Urabrelimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559232
 View more versions of this product
Product Code. 30284763
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Urabrelimab, also known as SRF231, is an investigational human monoclonal antibody targeting the CD47 protein, a cell surface antigen involved in immune evasion by cancer cells. The CD47 gene encodes a transmembrane glycoprotein broadly expressed on the surface of various cell types. Functionally, CD47 serves as a marker of self by binding to the Signal-regulatory protein alpha (SIRP alpha) receptor on myeloid cells, thereby preventing macrophage-mediated phagocytosis. This interaction is crucial in maintaining homeostasis by inhibiting the immune system from attacking healthy cells. However, CD47 is significantly overexpressed on the surface of many tumor cells, providing a mechanism for these cells to evade the immune system. As such, CD47 is a prominent target in cancer immunotherapy aimed at blocking this don't eat me signal to enhance the phagocytosis of cancer cells and improve patient prognosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Urabrelimab
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG4SP