missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£153.00 - £405.00
Specifications
| Antigen | ULK4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry |
| Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277431
|
Novus Biologicals
NBP1-20229-0.05ML |
0.05 mL |
£153.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231111
|
Novus Biologicals
NBP1-20229 |
0.15 mL |
£405.00
0.15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULK4 Polyclonal specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ULK4 | |
| Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse, Human | |
| DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans) | |
| ULK4 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| Q96C45 | |
| 54986 | |
| IgG | |
| Unpurified | |
| ULK4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title