missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-20229
This item is not returnable.
View return policy
Description
ULK4 Polyclonal specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ULK4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans) | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| ULK4. | |
| Human, Mouse | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry | |
| Q96C45 | |
| ULK4 | |
| A synthetic peptide from human ULK4 conjugated to blue carrier protein has been used as the antigen. The peptide shares 95% identity with mouse sequence. | |
| 0.15 mL | |
| Protein Kinase | |
| 54986 | |
| Reconstitute in 150 ul of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction