missing translation for 'onlineSavingsMsg'
Learn More

ULK4 Antibody, Novus Biologicals™

Product Code. 18277431 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.15 mL
Unit Size:
0.05mL
0.15mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18277431 0.05 mL 0.05mL
18231111 0.15 mL 0.15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18277431 Supplier Novus Biologicals Supplier No. NBP1202290.05ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

ULK4 Polyclonal specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK4
Applications Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunocytochemistry
Formulation Whole antisera with No Preservative
Gene Accession No. Q96C45
Gene Alias DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans)
Gene Symbols ULK4
Host Species Rabbit
Immunogen A synthetic peptide from the internal region of human ULK4 conjugated to an immunogenic carrier protein has been used as the immunogen. The antigen is located in aa region 750-800 of human ULK4, and shares 95% identity with the mouse sequence.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 54986
Test Specificity ULK4.
Reconstitution Reconstitute with 0.15 ml sterilized water.
Target Species Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.