missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE4A Recombinant Rabbit Monoclonal Antibody (24GB1785)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | UBE4A |
|---|---|
| Clone | 24GB1785 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275593
![]() |
Invitrogen™
MA557822 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275619
|
Invitrogen™
MA557823 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE4A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.Specifications
| UBE4A | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Ube4a | |
| 4732444G18Rik; 9930123J21Rik; Ab2-232; E4; KIAA0126; RING-type E3 ubiquitin transferase E4 A; UBE4A; Ubiquitin conjugation factor E4 A; ubiquitination factor E4A; ubiquitination factor E4A (UFD2 homolog, yeast); ubiquitination factor E4A, UFD2 homolog; UBOX2; UFD2; UFD2b | |
| Ube4a | |
| Primary | |
| Affinity Chromatography |
| 24GB1785 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9Q735, Q14139, Q6P7A2 | |
| 140630, 315608, 9354 | |
| A synthesized peptide derived from human UBE4A (aa 20-52). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title