missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE4A Recombinant Rabbit Monoclonal Antibody (24GB1785)

Description
UBE4A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.
Specifications
Specifications
| Antigen | UBE4A |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 24GB1785 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ube4a |
| Gene Accession No. | E9Q735, Q14139, Q6P7A2 |
| Gene Alias | 4732444G18Rik; 9930123J21Rik; Ab2-232; E4; KIAA0126; RING-type E3 ubiquitin transferase E4 A; UBE4A; Ubiquitin conjugation factor E4 A; ubiquitination factor E4A; ubiquitination factor E4A (UFD2 homolog, yeast); ubiquitination factor E4A, UFD2 homolog; UBOX2; UFD2; UFD2b |
| Gene Symbols | Ube4a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?