missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£271.00 - £433.00
Specifications
| Antigen | Tyrosine Hydroxylase |
|---|---|
| Dilution | Western Blot 1:50000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654618
|
Novus Biologicals
NBP3-05555-100ul |
100 μg |
£433.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608398
|
Novus Biologicals
NBP3-05555-50ul |
50 μg |
£271.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Tyrosine Hydroxylase | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Prostate Cancer, Tyrosine Kinases | |
| Supplied as an aliquot of IgY preparation | |
| 7054 | |
| IgY | |
| IgY purified |
| Western Blot 1:50000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Full length human Tyrosine Hydroxylase as expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title