Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05555-50ul
 View more versions of this product
Product Code. 18608398
  • £271.00 / 50µL
Description
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tyrosine Hydroxylase
Polyclonal
Western Blot 1:50000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:5000
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Full length human Tyrosine Hydroxylase as expressed in and purified from E. coli.
50 μg
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Prostate Cancer, Tyrosine Kinases
7054
Store at 4°C.
IgY
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Supplied as an aliquot of IgY preparation
Chicken
IgY purified
RUO
Primary
Human, Mouse, Rat
Purified