missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-05555-100ul
This item is not returnable.
View return policy
Description
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Tyrosine Hydroxylase | |
| Polyclonal | |
| Western Blot 1:50000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:5000 | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Full length human Tyrosine Hydroxylase as expressed in and purified from E. coli. | |
| 100 μg | |
| Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Prostate Cancer, Tyrosine Kinases | |
| 7054 | |
| Store at 4°C. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as an aliquot of IgY preparation | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction