missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 with One Hand Opening Mechanism
Safe and approved storage of flammable hazardous substances in working areas
£3031.60 - £3492.70
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Description | One-hand opening mechanism |
Dimensions (D x W x H) | 616 x 1196 x 1298 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
15905916
|
asecos™
3070700133753 |
2 x drawer (sheet steel powder-coated) | Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15995936
|
asecos™
3070700233753 |
2 x drawer (sheet steel powder-coated) | Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15915906
|
asecos™
3070700133744 |
2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15905936
|
asecos™
3070700233744 |
2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15915916
|
asecos™
3070700133754 |
3 x drawer (sheet steel powder-coated) | Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15905946
|
asecos™
3070700233754 |
3 x drawer (sheet steel powder-coated) | Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15925906
|
asecos™
3070700133745 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15915936
|
asecos™
3070700233745 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
616 x 1196 x 1298 mm | |
Steel | |
Wing | |
1298 mm | |
S90.129.120.WDEU | |
Powder-coated plain | |
1050 mm |
616 mm | |
One-hand opening mechanism | |
524 x 1050 x 1072 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1072 mm | |
2 | |
1196 mm |