missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 with One Hand Opening Mechanism
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3070700233754
Additional Details : Weight : 344.50919kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1072 mm | |
3 x drawer (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
524 mm | |
Steel | |
616 x 1196 x 1298 mm | |
524 x 1050 x 1072 mm | |
616 mm | |
Wing | |
2 |
One-hand opening mechanism | |
Safety storage cabinet | |
1050 mm | |
Powder-coated plain | |
1196 mm | |
359 kg | |
Yellow | |
1298 mm | |
S90.129.120.WDEU | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |