missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neville and More™ Type 3 Glass Injection Vials
Stores medical solutions or antibiotics.
£39.20 - £156.00
Specifications
Description | Glass injection vial with crimp neck |
---|---|
Color | Clear |
Material | Soda Lime Glass (Type 3) |
For Use With (Application) | Stores medical reagents or antibiotics |
Style | Cross section circular |
Product Code | Brand | Volume (Metric) Brim | Diameter (Metric) Inner Neck | Height (Metric) | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Brim | Diameter (Metric) Inner Neck | Height (Metric) | No. per Case | Price | Quantity & Availability | ||
15393418
|
Neville and More™
299684 |
50mL | 4.25cm | 7.3 cm (Neck) | 130 |
£156.00
Pack of 130 |
In Stock
Log in to see stock. |
||
15383418
|
Neville and More™
00770C3 |
10mL | 2.54cm | 5.35 m (Neck) | 168 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Features crimped neck
- Neck material: Pilfer proof glass
- Suited to contain liquids for various bottles, jars, laboratory consumables
- Tamper evident band
- Non reactive
- Material: Glass
Specifications
Glass injection vial with crimp neck | |
Soda Lime Glass (Type 3) | |
Cross section circular | |
2 cm |
Clear | |
Stores medical reagents or antibiotics | |
Crimp Cap |