missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neville and More™ Type 3 Glass Injection Vials
Stores medical solutions or antibiotics.
Brand: Neville and More™ 00770C3
Code : NEW
Additional Details : Weight : 3.19500kg
Description
- Features crimped neck
- Neck material: Pilfer proof glass
- Suited to contain liquids for various bottles, jars, laboratory consumables
- Tamper evident band
- Non reactive
- Material: Glass
Specifications
Glass injection vial with crimp neck | |
Clear | |
Stores medical reagents or antibiotics | |
Cross section circular | |
Crimp Cap | |
2 cm |
2.54cm | |
Soda Lime Glass (Type 3) | |
168 | |
10mL | |
5.35 m (Neck) |