missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neville and More™ Type 3 Glass Injection Vials
Stores medical solutions or antibiotics.
Brand: Neville and More™ 299684
Code : NEW
Additional Details : Weight : 6.27300kg
Description
- Features crimped neck
- Neck material: Pilfer proof glass
- Suited to contain liquids for various bottles, jars, laboratory consumables
- Tamper evident band
- Non reactive
- Material: Glass
Specifications
Glass injection vial with crimp neck | |
Clear | |
Stores medical reagents or antibiotics | |
Cross section circular | |
Crimp Cap | |
2 cm |
4.25cm | |
Soda Lime Glass (Type 3) | |
130 | |
50mL | |
7.3 cm (Neck) |