missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TXNRD1 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887973
|
Proteintech
11117-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877973
|
Proteintech
11117-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the family of pyridine nucleotide oxidoreductases. This protein reduces thioredoxins as well as other substrates, and plays a role in selenium metabolism and protection against oxidative stress. The functional enzyme is thought to be a homodimer which uses FAD as a cofactor. Each subunit contains a selenocysteine (Sec) residue which is required for catalytic activity. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenocysteine-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in several transcript variants encoding the same or different isoforms.Specifications
| TXNRD1 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| O89049, Q16881, Q9JMH6 | |
| 50493, 58819, 7296 | |
| TXNRD1 Fusion Protein Ag1618 | |
| Primary | |
| -20°C | |
| TXNRD1 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GRIM 12, GRIM12, KDRF, thioredoxin reductase 1, Thioredoxin reductase TR1, TR, TR1, TRXR1, TXNR, TXNRD1 | |
| Txnrd1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title