missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11117-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the family of pyridine nucleotide oxidoreductases. This protein reduces thioredoxins as well as other substrates, and plays a role in selenium metabolism and protection against oxidative stress. The functional enzyme is thought to be a homodimer which uses FAD as a cofactor. Each subunit contains a selenocysteine (Sec) residue which is required for catalytic activity. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenocysteine-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in several transcript variants encoding the same or different isoforms.Specifications
| TXNRD1 | |
| Polyclonal | |
| Unconjugated | |
| TXNRD1 | |
| GRIM 12, GRIM12, KDRF, thioredoxin reductase 1, Thioredoxin reductase TR1, TR, TR1, TRXR1, TXNR, TXNRD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50493, 58819, 7296 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O89049, Q16881, Q9JMH6 | |
| Txnrd1 | |
| TXNRD1 Fusion Protein Ag1618 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction