missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£167.00 - £360.00
Specifications
| Antigen | TXNIP |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-4ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18121325
|
Novus Biologicals
NBP2-27095SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729853
|
Novus Biologicals
NBP2-27095 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TXNIP Polyclonal specifically detects TXNIP in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot.Specifications
| TXNIP | |
| Western Blot 2-4ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1 | |
| TXNIP | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 94% homologous in mouse and rat. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Primate, Monkey | |
| Q9H3M7 | |
| 10628 | |
| A portion of amino acids 100-150 of human TxNIP was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title