missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-27095SS
This item is not returnable.
View return policy
Description
TXNIP Polyclonal specifically detects TXNIP in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot.
Specifications
| TXNIP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10628 | |
| Human, Mouse, Rat, Primate, Monkey | |
| Purified |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 2-4ug/ml | |
| Q9H3M7 | |
| TXNIP | |
| A portion of amino acids 100-150 of human TxNIP was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 94% homologous in mouse and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction