missing translation for 'onlineSavingsMsg'
Learn More

TXNIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NBP2-27095

 View more versions of this product

Product Code. 18729853

  • £360.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TXNIP Polyclonal specifically detects TXNIP in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TXNIP
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1
Rabbit
Protein A purified
RUO
10628
Human, Mouse, Rat, Primate, Monkey
Purified
Western Blot
1.0 mg/mL
Western Blot 2-4ug/ml
Q9H3M7
TXNIP
A portion of amino acids 100-150 of human TxNIP was used as the immunogen.
0.1 mg
Primary
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 94% homologous in mouse and rat.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only