missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TWEAK/TNFSF12 Antibody (RO5458640) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Clone | RO5458640 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045171
|
Novus Biologicals™
NBP3-28340-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044278
|
Novus Biologicals™
NBP3-28340-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047135
|
Novus Biologicals™
NBP3-28340-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| TWEAK/TNFSF12 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| TNFSF12/ TWEAK | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| RO5458640 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O43508 | |
| 8742 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title