Learn More
Novus Biologicals™ TWEAK/TNFSF12 Antibody (RO5458640) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28340-50ug
 View more versions of this product
Product Code. 30044278
  • £162.00 / 50µg
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TWEAK/TNFSF12
Monoclonal
Unconjugated
O43508
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
RO5458640
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
TNFSF12/ TWEAK
50 μg
Angiogenesis, Apoptosis, Cancer, Tumor Suppressors
8742
Human
Purified