missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TWEAK/TNFSF12 Antibody (RO5458640) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28340-100ug
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TWEAK/TNFSF12 | |
| Monoclonal | |
| Unconjugated | |
| O43508 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| RO5458640 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| TNFSF12/ TWEAK | |
| 100 μg | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| 8742 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction