missing translation for 'onlineSavingsMsg'
Learn More

TSPAN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TSPAN6
Concentration 0.13 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16839923
Dokumentvisning
Proteintech
12293-1-AP-20UL
20 μL
£135.00
20µL
Se alternative produkter
Spar op til 
»
Please to purchase this item. Need a web account? Register with us today!
16829923
Dokumentvisning
Proteintech
12293-1-AP-150UL
150 μL
£349.00
150µL
Se alternative produkter
Spar op til 
»
Please to purchase this item. Need a web account? Register with us today!
Beskrivelse

Beskrivelse

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.
Tekniske data

Tekniske data

TSPAN6
Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
O43657, O70401
302313, 56496, 7105
TSPAN6 Fusion Protein Ag2964
Primary
-20°C
TSPAN6
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A15 homolog, T245, T245 protein, tetraspanin 6, Tetraspanin TM4 D, TM4SF6, TSPAN 6, TSPAN6
Tspan6
IgG
Antigen Affinity Chromatography
Antibody
Videoer
SDS
Dokumenter

Dokumenter

Produktcertifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.