missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12293-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.Specifications
| TSPAN6 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN6 | |
| A15 homolog, T245, T245 protein, tetraspanin 6, Tetraspanin TM4 D, TM4SF6, TSPAN 6, TSPAN6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 302313, 56496, 7105 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43657, O70401 | |
| Tspan6 | |
| TSPAN6 Fusion Protein Ag2964 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction