Learn More
TSPAN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12293-1-AP-150UL
 View more versions of this product
Product Code. 16829923
  • £349.00 / 150µL
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.
Specifications
Specifications
TSPAN6
Polyclonal
Unconjugated
TSPAN6
A15 homolog, T245, T245 protein, tetraspanin 6, Tetraspanin TM4 D, TM4SF6, TSPAN 6, TSPAN6
Rabbit
Antigen Affinity Chromatography
RUO
302313, 56496, 7105
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43657, O70401
Tspan6
TSPAN6 Fusion Protein Ag2964
150 μL
Primary
Human, Mouse, Rat