Learn More
Transglutaminase II Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
£121.38 - £693.60
Specifications
Antigen | Transglutaminase II Ab-1 |
---|---|
Clone | CUB 7402 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12630597
|
Epredia
MS-224-B1 |
Concentrated | Biotin | 500 μL |
£490.00
500µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12660597
|
Epredia
MS-224-P1 |
Concentrated | Unconjugated | 500 μL |
£374.00
500µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12640597
|
Epredia
MS-224-P |
Concentrated | Unconjugated | 1 mL |
£626.00
1mL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12650597
|
Epredia
MS-224-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12670597
|
Epredia
MS-224-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12690597
|
Epredia
MS-224-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12620597
|
Epredia
MS-224-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12610597
|
Epredia
MS-224-B |
Concentrated | Biotin | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12620607
|
Epredia
MS-224-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.Host Species: Mouse
Clone: CUB 7402
Isotype: IgG1/κ
Species Reactivity: Human, Dog, Rabbit and Guinea Pig. Does not react with mouse and rat. Others not known.
Epitope: aa 447-478
Immunogen: Purified guinea pig liver TGase II
Molecular Weight: 77-85kDa
Positive Control: HUVEC cells, Breast CA
Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-1 inhibits the TGase II activity.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Affinity Purification
- Biological Blockade
Note:
USA: RUO; Int'l: RUO
Specifications
Transglutaminase II Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Mouse | |
Neuroscience | |
Purified guinea pig liver TGase II | |
Primary |
CUB 7402 | |
Monoclonal | |
RUO | |
Canine, Guinea Pig, Human, Rabbit | |
IgG2a κ |