Learn More
Invitrogen™ TRAFD1 Monoclonal Antibody (K52015_1F2), Invitrogen™
Mouse Monoclonal Antibody
£453.00 - £760.00
Specifications
| Antigen | TRAFD1 |
|---|---|
| Clone | K52015_1F2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30208736
|
Invitrogen™
MA556232 |
200 μL |
£760.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211212
|
Invitrogen™
MA555852 |
50 μL |
£453.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RLCDNCKKEI PVFNFTIHEI HCQRNIGMCP TCKEPFPKSD METHMAAEHC QVTCKCNKKL EKRLLKKHEE TECPLRLAVC QHCDLELSIL KLKEHEDYCG ARTELCGNCG RNVLVKDLKT HPEVCGREGE EKRNEVAIPP NAYDESWGQD GIWIASQLLR QIEALDPPMR LPRRPLRAFE SDVFHNRTTN QRNITAQVSI QNNLFEEQER QERNRGQQPP KEGGEESANL DFMLALSLQN EGQASSVAEQ DFWRAVCEAD QSHGGPRSLS DIKGAADEIM LPCEFCEELY PEELLIDHQT SCNPSRALPS LNTGSSSPRG VEEPDVIFQN
TRAFD1 Monoclonal antibody specifically detects TRAFD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| TRAFD1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1 | |
| Recombinant human TRAFD1 (immunogen range is 11-340). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TRAFD1 |
| K52015_1F2 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14545 | |
| 10906 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.