Learn More
Invitrogen™ TRAFD1 Monoclonal Antibody (K52015_1F2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555852
Description
Sequence of this protein is as follows: RLCDNCKKEI PVFNFTIHEI HCQRNIGMCP TCKEPFPKSD METHMAAEHC QVTCKCNKKL EKRLLKKHEE TECPLRLAVC QHCDLELSIL KLKEHEDYCG ARTELCGNCG RNVLVKDLKT HPEVCGREGE EKRNEVAIPP NAYDESWGQD GIWIASQLLR QIEALDPPMR LPRRPLRAFE SDVFHNRTTN QRNITAQVSI QNNLFEEQER QERNRGQQPP KEGGEESANL DFMLALSLQN EGQASSVAEQ DFWRAVCEAD QSHGGPRSLS DIKGAADEIM LPCEFCEELY PEELLIDHQT SCNPSRALPS LNTGSSSPRG VEEPDVIFQN
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
Specifications
| TRAFD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O14545 | |
| TRAFD1 | |
| Recombinant human TRAFD1 (immunogen range is 11-340). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K52015_1F2 | |
| Unconjugated | |
| TRAFD1 | |
| 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10906 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.