missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAFD1 Monoclonal Antibody (K52015_1F2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556232

 View more versions of this product

Product Code. 30208736

  • £760.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: RLCDNCKKEI PVFNFTIHEI HCQRNIGMCP TCKEPFPKSD METHMAAEHC QVTCKCNKKL EKRLLKKHEE TECPLRLAVC QHCDLELSIL KLKEHEDYCG ARTELCGNCG RNVLVKDLKT HPEVCGREGE EKRNEVAIPP NAYDESWGQD GIWIASQLLR QIEALDPPMR LPRRPLRAFE SDVFHNRTTN QRNITAQVSI QNNLFEEQER QERNRGQQPP KEGGEESANL DFMLALSLQN EGQASSVAEQ DFWRAVCEAD QSHGGPRSLS DIKGAADEIM LPCEFCEELY PEELLIDHQT SCNPSRALPS LNTGSSSPRG VEEPDVIFQN

The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRAFD1
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O14545
TRAFD1
Recombinant human TRAFD1 (immunogen range is 11-340).
200 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K52015_1F2
Unconjugated
TRAFD1
1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1
Mouse
Protein A
RUO
10906
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.