missing translation for 'onlineSavingsMsg'
Learn More

TIP47 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £375.00

Specifications

Antigen TIP47
Applications Immunofluorescence
Classification Polyclonal
Conjugate CoraLite 555
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17761339
View Documents
Proteintech
CL555-10694-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17649896
View Documents
Proteintech
CL555-10694-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIP47 Polyclonal antibody apecifically detetects TIP47 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

TIP47
Polyclonal
Liquid
Human
BC007566
M6PRBP1
IgG
Antigen affinity purification
Immunofluorescence
CoraLite 555
Rabbit
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
10226
TIP47 fusion protein Ag1028
Primary
Store at -20°C. Avoid exposure to light.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.