missing translation for 'onlineSavingsMsg'
Learn More

TIP47 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL555-10694-20UL

 View more versions of this product

Product Code. 17761339

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIP47 Polyclonal antibody apecifically detetects TIP47 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

TIP47
Polyclonal
CoraLite 555
BC007566
Rabbit
Antigen affinity purification
Primary
Human
Liquid
Immunofluorescence
1000 ug/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
M6PRBP1
TIP47 fusion protein Ag1028
20 μL
10226
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.