missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP47 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL555-10694-100UL
This item is not returnable.
View return policy
Description
TIP47 Polyclonal antibody apecifically detetects TIP47 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| TIP47 | |
| Polyclonal | |
| CoraLite 555 | |
| BC007566 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| M6PRBP1 | |
| TIP47 fusion protein Ag1028 | |
| 100 μL | |
| 10226 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction