missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thyroglobulin Monoclonal Antibody (TG8.1), Invitrogen™

Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | Thyroglobulin |
|---|---|
| Clone | TG8.1 |
| Concentration | 1.2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17860416
|
Invitrogen™
MA5-46142 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11558882
![]() |
Invitrogen™
MA110595 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17850466
|
Invitrogen™
MA5-45993 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10595 detects Thyroglobulin, specifically both T3 and T4, from human samples. MA1-10595 has been successfully used in ELISA applications. The MA1-10595 immunogen is purified human Thyroglobulin.
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.Specifications
| Thyroglobulin | |
| 1.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor | |
| TG | |
| IgM | |
| Protein A | |
| Antibody |
| TG8.1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01266 | |
| 7038 | |
| Purified human Thyroglobulin. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| TG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title