missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thyroglobulin Monoclonal Antibody (TG8.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-45993
This item is not returnable.
View return policy
Description
MA1-10595 detects Thyroglobulin, specifically both T3 and T4, from human samples. MA1-10595 has been successfully used in ELISA applications. The MA1-10595 immunogen is purified human Thyroglobulin.
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
Specifications
| Thyroglobulin | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P01266 | |
| TG | |
| Purified human Thyroglobulin. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| ELISA | |
| TG8.1 | |
| Unconjugated | |
| TG | |
| AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor | |
| Mouse | |
| Protein A | |
| RUO | |
| 7038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction