Learn More
Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.
£156.06 - £591.60
Specifications
Antigen | Thrombospondin (TSP) Ab-11 |
---|---|
Clone | D4.6 + A6.1 + MBC 200.1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12648806
|
Epredia
MS-1066-P1 |
200μg/mL | 500 μL |
£410.00
500µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12638806
|
Epredia
MS-1066-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12658806
|
Epredia
MS-1066-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12668806
|
Epredia
MS-1066-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Host Species: Mouse
Clone: D4.6 + A6.1 + MBC 200.1
Isotype: IgG1 + IgG1 + IgG1
Species Reactivity: Human. Others not known.
Immunogen: Purified human TSP
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting (Reduced and nonreduced)
Note:
USA: RUO; Int'l: RUO
Specifications
Thrombospondin (TSP) Ab-11 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG1 |
D4.6 + A6.1 + MBC 200.1 | |
Monoclonal | |
Mouse | |
Angiogenesis | |
Purified human TSP | |
Primary |