Learn More
Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.
Brand: Epredia MS-1066-P0
Code : LD
Additional Details : Weight : 1.00000kg
Description
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Host Species: Mouse
Clone: D4.6 + A6.1 + MBC 200.1
Isotype: IgG1 + IgG1 + IgG1
Species Reactivity: Human. Others not known.
Immunogen: Purified human TSP
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting (Reduced and nonreduced)
Note:
USA: RUO; Int'l: RUO
Specifications
Thrombospondin (TSP) Ab-11 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Angiogenesis | |
Human | |
IgG1 |
Western Blot | |
D4.6 + A6.1 + MBC 200.1 | |
Unconjugated | |
Purified human TSP | |
RUO | |
Primary | |
Concentrated |