missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 (Transcription Factor 4) Mouse, Clone: TCF4/1705, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | TCF4 (Transcription Factor 4) |
|---|---|
| Clone | TCF4/1705 |
| Concentration | 200μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| TCF4 (Transcription Factor 4) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human TCF4 protein fragment (aa365-671) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 71kDa |
| TCF4/1705 | |
| ELISA, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 6925 | |
| IgG2a κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear | |
| HeLa or HepG2 cells. Bladder |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title