missing translation for 'onlineSavingsMsg'
Learn More

TCF4 (Transcription Factor 4) Mouse, Clone: TCF4/1705, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 6925-MSM1-P1

 View more versions of this product

Product Code. 15930938

  • £423.29 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TCF4 (Transcription Factor 4)
Nuclear
TCF4/1705
Unconjugated
Mouse
71kDa
Protein A/G purified from Bioreactor concentrate.
RUO
6925
IgG2a κ
ELISA, Western Blot, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human TCF4 protein fragment (aa365-671)
HeLa or HepG2 cells. Bladder
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.