missing translation for 'onlineSavingsMsg'
Learn More

TCF4 (Transcription Factor 4) Mouse, Clone: TCF4/1705, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 6925-MSM1-P0

 View more versions of this product

Product Code. 15920938

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TCF4 (Transcription Factor 4)
Nuclear
TCF4/1705
Unconjugated
Mouse
71kDa
Protein A/G purified from Bioreactor concentrate.
RUO
6925
IgG2a κ
ELISA, Western Blot, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human TCF4 protein fragment (aa365-671)
HeLa or HepG2 cells. Bladder
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.