missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ AADvanced™ Dead Cell Stain Kit

SYTOX™ AADvanced™ Dead Cell Stain is a new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry.

Brand:  Molecular Probes™ S10274

Code : 50

Additional Details : Weight : 0.00450kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II

 View more versions of this product

Product Code. 10277064

  • £306.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • A new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry
  • The dye is spectrally similar to 7-AAD but with rapid kinetics of uptake and relatively low CVs
  • SYTOX™ AADvanced™ Dead Cell Stain penetrates cells more efficiently than 7-AAD providing better separation of live and dead cells
  • It can also be used with fixed cells for DNA content analysis when paired with RNAse treatment
  • Kits contain a vial(s) of dried dye and anhydrous DMSO providing stable shelf life
Specifications

Specifications

Impermeant
Contains 5 vials of SYTOX™ AADvanced™ dead cell stain and 500 μL dimethylsulfoxide (DMSO).

Store at ≤-20°C and protect from light.
Fluorescence
Flow Cytometer
SYTOX™ AADvanced™
546 nm
Tube(s)
500 Reactions
DMSO (Dimethylsulfoxide)
Red
SYTOX™ AADvanced™ Dead Cell Stain Kit
Viability Assay
Dead Cell Stain Kit
647 nm
Solid
Molecular Probes™, SYTOX™
Room Temperature
Nucleus, Nucleic Acids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.