missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ SYTOX™ AADvanced™ Dead Cell Stain Kit
SYTOX™ AADvanced™ Dead Cell Stain is a new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry.
£128.00 - £306.00
Specifications
Cell Permeability | Impermeant |
---|---|
Color | Red |
Description | SYTOX™ AADvanced™ Dead Cell Stain Kit |
Detection Method | Fluorescence |
For Use With (Application) | Viability Assay |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10287064
|
Molecular Probes™
S10349 |
100 Reactions |
£128.00
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
10277064
|
Molecular Probes™
S10274 |
500 Reactions |
£306.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry
- The dye is spectrally similar to 7-AAD but with rapid kinetics of uptake and relatively low CVs
- SYTOX™ AADvanced™ Dead Cell Stain penetrates cells more efficiently than 7-AAD providing better separation of live and dead cells
- It can also be used with fixed cells for DNA content analysis when paired with RNAse treatment
- Kits contain a vial(s) of dried dye and anhydrous DMSO providing stable shelf life
Specifications
Impermeant | |
SYTOX™ AADvanced™ Dead Cell Stain Kit | |
Viability Assay | |
Dead Cell Stain Kit | |
647 nm | |
Solid | |
Molecular Probes™, SYTOX™ | |
DMSO (Dimethylsulfoxide) |
Red | |
Fluorescence | |
Flow Cytometer | |
SYTOX™ AADvanced™ | |
546 nm | |
Tube(s) | |
Room Temperature | |
Nucleus, Nucleic Acids |
For Research Use Only. Not for use in diagnostic procedures.