missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00 - £397.00
Specifications
| Antigen | SYT17 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244134
|
Novus Biologicals
NBP1-03494-0.1MG |
0.1 mg |
£141.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18261540
|
Novus Biologicals
NBP1-03494 |
0.5 mg |
£397.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYT17 Polyclonal antibody specifically detects SYT17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen).Specifications
| SYT17 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Q9BSW7, Q920M7, Q62807 | |
| 51760 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| B/K protein, synaptotagmin XVIIProtein B/K, synaptotagmin-17, sytXVII | |
| SYT17 | |
| Primary | |
| SYT17. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title