missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03494
This item is not returnable.
View return policy
Description
SYT17 Polyclonal antibody specifically detects SYT17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications
| SYT17 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS with No Preservative | |
| B/K protein, synaptotagmin XVIIProtein B/K, synaptotagmin-17, sytXVII | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51760 | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
| Q9BSW7, Q920M7, Q62807 | |
| SYT17 | |
| A synthetic peptide from the internal region of human SYT17 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in rat and mouse. | |
| 0.5 mg | |
| Primary | |
| SYT17. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction