Learn More
SYT17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-03494-0.1MG
 View more versions of this product
Product Code. 18244134
  • £141.00 / 0.10mg
Description
SYT17 Polyclonal specifically detects SYT17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SYT17
Polyclonal
Unconjugated
PBS with No Preservative
B/K protein, synaptotagmin XVIIProtein B/K, synaptotagmin-17, sytXVII
Rabbit
IgG purified
RUO
51760
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
LYOPH
Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 10-50 ug/ml
Q9BSW7, Q920M7, Q62807
SYT17
A synthetic peptide from the internal part of human Synaptotagmin 17 (SYT17) conjugated to an immunogenic carrier protein was used as the immunogen.
0.1 mg
Primary
SYT17.
Human
Purified