missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntrophin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | Syntrophin |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18337481
|
Bio-Techne
NBP3-17941-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342604
|
Novus Biologicals
NBP3-17941-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntrophin Polyclonal antibody specifically detects Syntrophin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Syntrophin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GTRHGVDTHLFSVESPQELAAWTRQLVDGCHRAAEGVQEVSTACTWNGRPCSLSVHIDKGFTLWAAEPGAARAVLLRQPFEKLQMS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 6640 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title