Learn More
SV40 Large T Antigen Ab-2 Mouse Monoclonal Antibody, BSA and azide, Lab Vision™ Epredia™
Epredia™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
£105.00 - £520.20
Specifications
Antigen | SV40 Large T Antigen Ab-2 |
---|---|
Clone | PAb108 |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12660237
|
Epredia
MS-1833-P1 |
200μg/mL | 500 μL |
£344.00
500µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12650237
|
Epredia
MS-1833-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12640237
|
Epredia
MS-1833-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12670237
|
Epredia
MS-1833-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12680237
|
Epredia
MS-1833-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.Host Species: Mouse
Clone: PAb108
Isotype: IgG2a1
Species Reactivity: SV40 virus.
Epitope: N-terminal
Immunogen: SV40-transformed mouse line B4
Molecular Weight: 94kDa
Positive Control: SV40-transfected cells
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
SV40 Large T Antigen Ab-2 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Infectious Disease | |
SV40 transformed mouse line B4 |
PAb108 | |
Monoclonal | |
Concentrated | |
RUO | |
Simian virus 40 (SV40) | |
Primary |