missing translation for 'onlineSavingsMsg'
Learn More

SV40 Large T Antigen Ab-2 Mouse Monoclonal Antibody, BSA and azide, Lab Vision™ Epredia™

Epredia™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-1833-PABX

Code : LD

Additional Details : Weight : 0.00180kg

 View more versions of this product

Product Code. 12680237

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb108

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: N-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

SV40 Large T Antigen Ab-2
Monoclonal
1mg/mL
Mouse
200 μL
Infectious Disease
Simian virus 40 (SV40)
Immunofluorescence, Immunoprecipitation, Western Blot
PAb108
Unconjugated
SV40 transformed mouse line B4
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers