missing translation for 'onlineSavingsMsg'
Learn More

SV40 Large T Antigen Ab-2 Mouse Monoclonal Antibody, BSA and azide, Lab Vision™ Epredia™

Epredia™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-1833-P1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12660237

  • £344.00 / 500µL
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb108

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: N-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

SV40 Large T Antigen Ab-2
Monoclonal
200μg/mL
Mouse
500 μL
Infectious Disease
Simian virus 40 (SV40)
Immunofluorescence, Immunoprecipitation, Western Blot
PAb108
Unconjugated
SV40 transformed mouse line B4
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers