missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420H2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £367.00
Specifications
| Antigen | SUV420H2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Knockout Validated |
| Applications | Western Blot, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605302
|
Novus Biologicals
NBP2-94300-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690502
|
Novus Biologicals
NBP2-94300-0.1ml |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV420H2 Polyclonal antibody specifically detects SUV420H2 in Human, Mouse samples. It is validated for Western Blot, Gene Knock-OutSpecifications
| SUV420H2 | |
| Western Blot, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human KMT5C (NP_116090.2). FLPESGFTILPCTRYSMETNGAKIVSTRAWKKNEKLELLVGCIAELREADEGLLRAGENDFSIMYSTRKRSAQLWLGPAAFINHDCKPNCKFVPADGNAAC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 84787 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title